Would you like to log in again?
HiTrap Columns, Q XL, Cytiva
Supplier: Cytiva
HiTrap™
17-5159-01 17-5158-01 17-5159-01 17-5158-01
95056-108PK 204.75 USD InStock
95056-108 CA95056-108L CA95056-110L 95056-110
HiTrap Columns, Q XL, Cytiva
Chromatography Columns
HiTrap Q XL are strong anion exchangers prepacked with Q Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography.
  • High binding capacities with high flow rates
  • Convenient and affordable for fast, easy separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump, or chromatography system such as ÄKTA design
Q Sepharose XL and SP Sepharose XL resins have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chains increase the exposure of the Q or SP charged groups, which results in very high loading capacities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty