Would you like to log in again?
HiTrap Columns, SP XL, Cytiva
Supplier: Cytiva
HiTrap™
17-5161-01 17-5160-01 17-5161-01 17-5160-01
95056-114PK 461.24 USD InStock
CA95056-114L 95056-114 CA95056-112L 95056-112
HiTrap Columns, SP XL, Cytiva
Chromatography Columns
HiTrap SP XL are strong cation exchangers prepacked with SP Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography.
  • High binding capacities with high flow rates
  • Convenient and affordable for fast, easy separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump, or chromatography system such as ÄKTA design
Q Sepharose XL and SP Sepharose XL resins have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chains increase the exposure of the Q or SP charged groups, which results in very high loading capacities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty