Would you like to log in again?
HiTrap Columns, Chelating HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0409-01 17-0409-03 17-0409-01 17-0408-01 17-0408-01 17-0409-03
95056-038PK 262.94 USD InStock
CA95056-038L CA95056-226L 95056-038 95056-226 95056-144 CA95056-144L
HiTrap Columns, Chelating HP, Cytiva
Chromatography Columns
HiTrap Chelating HP is prepacked with Chelating Sepharose High Performance. The medium is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.
  • For optimizing purification of histidine-tagged proteins using different kinds of metal ions
  • Conveniently charge with your metal of choice
  • Iminidoacetic acid coupled to Sepharose High Performance
  • Purifying proteins with exposed histidine groups via metal ion complex formation
  • Simple operation with a syringe, pump or chromatographic system such as ÄKTA design or FPLC System
HiTrap Chelating HP is a prepacked ready to use, column for preparative affinity chromatography. The special design of the column, together with the matrix, provide fast, simple and easy separations in a convenient format.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty