Would you like to log in again?
HiTrap Columns, MabSelect Xtra, Cytiva
Supplier: Cytiva
HiTrap™
28-4082-60 28-4082-58 28-4082-58 28-4082-61 28-4082-60 28-4082-61
95056-162PK 1020.41 USD InStock
95056-162 CA95056-234L CA95055-972L CA95056-162L 95055-972 95056-234
HiTrap Columns, MabSelect Xtra, Cytiva
Chromatography Columns
HiTrap MabSelect Xtra columns are prepacked with MabSelect Xtra, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.
  • Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect medium
  • MabSelect Xtra uses the same recombinant protein A ligand as MabSelect
  • Oriented coupling of recombinant Protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs
The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty