Would you like to log in again?
Vivaspin Sample Concentrators, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VivaSpin™
28-9323-17 28-9322-94 28-9322-47 28-9322-37 28-9322-58 28-9322-35 28-9322-25 28-9322-36 28-9322-47 28-9322-45 28-9323-61 28-9322-48 28-9322-40 28-9323-62 28-9322-96 28-9323-18 28-9322-45 28-9323-58 28-9323-63 28-9323-58 28-9322-18 28-9323-19 28-9323-59 28-9323-61 28-9322-23 28-9323-60 28-9322-57 28-9322-96 28-9323-19 28-9323-18 28-9323-59 28-9322-40 28-9322-35 28-9322-57 28-9322-36 28-9323-62 28-9322-93 28-9323-63 28-9322-23 28-9322-37 28-9322-94 28-9322-48 28-9322-93 28-9322-18 28-9322-25 28-9323-60 28-9323-17 28-9322-58
95055-990PK 215.32 USD InStock
95055-990 CA95056-124L CA95056-126L CA95056-128L 95055-832 95055-996 95055-830 95055-994 95055-992 CA95055-832L 95055-828 CA95055-830L CA95055-836L 95056-180 95055-826 CA95055-834L 95056-182 CA95056-180L 95055-988 95056-126 CA95056-172L CA95056-182L CA95055-828L CA95056-130L 95056-128 CA95056-174L CA95055-826L CA95056-176L CA95056-132L 95056-124 CA95056-178L CA95056-134L CA95055-992L CA95055-990L CA95055-996L 95056-172 CA95055-994L 95056-130 95056-174 95055-914 95055-836 CA95055-988L 95055-834 CA95055-914L 95056-132 95056-176 95056-178 95056-134
Vivaspin Sample Concentrators, Cytiva
Filters Centrifugal Filters
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration.
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss
  • Dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • Vertical PES membrane minimises membrane blockage and tolerates high flow rates
  • Compatible pH range is from pH 1 to 9
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cutoff (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3000 to 100000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty