Would you like to log in again?
RESOURCE ISO Columns, Cytiva
Supplier: Cytiva
17-1185-01 17-1185-01
95056-194EA 1433.44 USD InStock
95056-194 CA95056-194L
RESOURCE ISO Columns, Cytiva
Chromatography Columns
RESOURCE ISO are prepacked SOURCE 15 ISO (isopropyl) columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC). For fast, high-resolution purifications, RESOURCE columns are excellent choice for polishing and removal of trace impurities.
  • High performance at low back pressure, even with high sample load of up to 25 mg of protein
  • Hydrophobic ligand (isopropyl) coupled to the 15 µm monodispersed rigid polystyrene/divinylbenzene matrix
  • Designed for low- to medium- pressure systems ranging from a peristalic pump to ÄKTA design and FPLC Systems
RESOURCE HIC columns are available with ether, isopropyl, or phenyl. Generally, RESOURCE PHE will have the strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty