Would you like to log in again?
HiTrap Columns, Benzamidine FF (High Sub), Cytiva
Supplier: Cytiva
HiTrap™
17-5144-01 17-5144-01 17-5143-02 17-5143-01 17-5143-02 17-5143-01
95056-098EA 264.06 USD InStock
95056-098 CA95056-198L CA95056-098L 95056-198 CA95056-200L 95056-200
HiTrap Columns, Benzamidine FF (High Sub), Cytiva
Chromatography Columns
HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
  • High binding capacity
  • It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system
HiTrap Benzamidine FF (high sub) are prepacked 1 and 5 ml columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
The medium, Benzamidine Sepharose 4 Fast Flow (high sub), is also available as lab packages and is an excellent choice for scale-up.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty