Would you like to log in again?
HiTrap Columns, Capto Q, Cytiva
Supplier: Cytiva
HiTrap™
11-0013-02 11-0013-03 11-0013-02 11-0013-03
95056-218PK 271.37 USD InStock
95056-218 95055-922 CA95056-218L CA95055-922L
HiTrap Columns, Capto Q, Cytiva
Chromatography Columns
HiTrap Capto Q chromatography column is prepacked with a strong anion exchange modern resin for small scale protein purification.
  • Suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications
  • Quaternary ammonium (Q) strong anion exchanger
  • Optimized for capture and intermediate protein purification
  • Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
Capto Q resin couples a quaternary ammonium (Q) strong anion exchanger to a chemically modified, high-flow agarose matrix. The rigid agarose matrix provides greater particle rigidity without compromising pore size, which allows for high bed volume and purification of viscous samples at high flow rates.
High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix. The resin’s high binding-capacity, high flow rate, and low back pressure reduce process cycle times and improve productivity at process scale.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty