Would you like to log in again?
PD MidiTrap G-25, Cytiva
Supplier: Cytiva
MidiTrap™
28-9180-08 28-9180-08
95056-250PK 575.66 USD InStock
95056-250 CA95056-250L
PD MidiTrap G-25, Cytiva
Chromatography Columns
PD MidiTrap G-25 is convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, The use of centrifugation protocol with all gravity columns.
  • Reliable and reproducible clean-up of proteins and other large biomolecules (>5000 molecular weight)
  • Ideal for desalting, buffer exchange, and removal of low molecular weight compounds
  • High desalting capacity
  • Flexible protocols to use with gravity flow or centrifugation
  • Sample preparation in the range of 0,5 to 1,0 ml for gravity and 0,75 to 1,0 ml with centrifugation
PD MidiTrap G-25 is designed for convenient desalting and buffer exchange of biological samples and volumes up to 1.0 ml can be handled. Two alternative application protocols are available, gravity or centrifugation. By using the gravity protocol, a simple cleanup of the sample is done with one or several columns in parallel without any need for a purification system. With the centrifugation protocol, the samples are run in parallel in a standard centrifuge with minimal dilution of the eluted sample.
For larger scale please use PD-10 desalting column (2,5 ml) and for smaller scale, PD MiniTrap™ G-25 (0,5 ml), PD SpinTrap™ G-25 (130 µl) and PD MultiTrap™ G-25 (130 µl) in parallel.
Delivery information: Includes four adapters to enable easy centrifugation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty