Would you like to log in again?
PD MiniTrap G-10 Columns, Cytiva
Supplier: Cytiva
MidiTrap™ MiniTrap™
28-9180-10 28-9180-10
95056-252PK 597.33 USD InStock
95056-252 CA95056-252L
PD MiniTrap G-10 Columns, Cytiva
Chromatography Columns
PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.
  • Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700 before downstream analysis such as liquid chromatography, electrophoresis, and MS
  • Efficient removal of contaminants such as salts, dyes, and radioactive labels
  • High desalting capacity
  • Preparation of samples in the volume range of 100 to 300 μl
PD MiniTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system. PD MiniTrap G-10 columns are a member of the Trap platform, which addresses the need for flexible, small-scale preparation of proteins and peptides before analysis.
For larger scale please use PD MidiTrap G-10 columns.
Delivery information: Instructions are supplied with the columns.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty