Would you like to log in again?
HiTrap Columns, Phenyl FF (High Sub), Cytiva
Supplier: Cytiva
HiTrap™
17-1355-01 17-5193-01 17-5193-01 17-1355-01
95055-818PK 658.21 USD InStock
95055-818 CA95056-258L CA95055-818L 95056-258
HiTrap Columns, Phenyl FF (High Sub), Cytiva
Chromatography Columns
Phenyl Sepharose 6 Fast Flow is a well-established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.
  • Provide convenient and fast small-scale protein purifications
  • Two levels of substitution on Phenyl Sepahrose 6 Fast Flow help find optimal selectivity and binding capacity
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
HiTrap Phenyl FF (high sub) are prepacked Phenyl Sepharose 6 Fast Flow columns for hydrophobic interaction chromatography (HIC). The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty