Would you like to log in again?
HiTrap Columns, Butyl FF, Cytiva
Supplier: Cytiva
HiTrap™
17-5197-01 17-1357-01 17-1357-01 17-5197-01
95056-260PK 302.47 USD InStock
95056-260 CA95055-824L 95055-824 CA95056-260L
HiTrap Columns, Butyl FF, Cytiva
Chromatography Columns
HiTrap Butyl FF are prepacked Butyl Sepharose 4 Fast Flow columns for hydrophobic interaction chromatography (HIC).
  • For screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Provide convenient and fast small-scale protein purifications
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions. Butyl Sepharose 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty