Would you like to log in again?
GST MultiTrap Filter Plates, Cytiva
Supplier: Cytiva
Multitrap™
28-4055-00 28-4055-00
95056-294PK 1296.92 USD InStock
95056-294 CA95056-294L
GST MultiTrap Filter Plates, Cytiva
Microplates Filter Plates
GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and purification of glutathione S-transferase (GST) tagged proteins
  • 96-well filter plate format for high-throughput screening and parallel purification of GST-tagged proteins
  • Highly reproducible well-to well and plate-to-plate results
  • High purity in one step, high chemical stability
  • Load unclarified samples directly to the wells
  • Wells are prepacked with Glutathione Sepharose media for convenience
  • Easy and predictable scale-up using prepacked GSTrap FF, GSTPrep FF 16/10, and GSTrap 4B columns
Purification of up to 0.5 mg of GST-tagged proteins/well directly from unclarified cell lysate can be achieved using GST MultiTrap filter plates. Direct purification of GST-tagged proteins from unclarified lysate shortens handling time and minimizes degradation of sensitive target proteins. The 96-well plate format gives great flexibility, both when working with automated robotic systems and manually using centrifugation or vacuum. Consistent well-to-well and plate-to-plate performance ensures high reproducibility.
Centrifugation force max.: 700 xg
Delivery information: GST MultiTrap FF is prepacked with Glutathione Sepharose 4 fast flow, while GST MultiTrap 4B is prepacked with Glutathione Sepharose 4B.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty