Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Immobiline™ DryStrip Gels, Cytiva
Supplier: Cytiva
Immobiline™
17-6002-47 17-6001-12 17-6001-97 18-1016-61 17-6003-76 17-6003-74 17-1233-01 17-6002-46 17-6001-95 18-1016-60 17-1234-01 17-6002-46 17-6001-14 17-6001-94 17-6003-73 17-6003-74 17-1234-01 17-6001-95 17-6001-12 17-6001-94 17-1235-01 17-6001-96 18-1016-60 17-6003-75 17-6001-13 17-6001-14 17-6001-11 17-6001-11 17-6001-15 17-6001-10 17-6002-47 17-6001-13 17-6002-44 18-1016-61 17-6001-97 17-6002-44 17-6002-45 17-1235-01 17-6002-45 17-6003-77 17-6001-15 17-6003-73 17-6001-96 17-6003-75 17-1233-01 17-6003-76 17-6001-10 17-6003-77
95056-416PK 222.23 USD InStock
CA95056-366L CA95056-552L CA95056-378L CA95056-368L 95056-416 95056-438 95056-418 95056-382 95056-384 95056-362 95056-380 95056-368 CA95056-380L CA95056-548L 95056-546 CA95056-382L CA95056-546L 95056-402 CA95056-384L CA95056-362L 95056-364 95056-440 CA95056-554L 95056-366 CA95056-364L 95056-420 CA95056-410L 95056-408 CA95056-412L CA95056-402L 95056-548 CA95056-414L 95056-404 CA95056-404L CA95056-438L CA95056-416L CA95056-406L 95056-406 CA95056-418L CA95056-408L 95056-412 95056-554 95056-378 95056-414 CA95056-440L 95056-410 CA95056-420L 95056-552
Immobiline™ DryStrip Gels, Cytiva
Electrophoresis Gels Precast Gels
Immobiline™ DryStrip gels (IPG strips) are used for isoelectric focusing (IEF), run as the first dimension of 2-D electrophoresis or as a separate application. IPG strips contain a preformed pH gradient immobilised in precast polyacrylamide gels on a plastic backing. A comprehensive range of overlapping IPG strips covering narrow, medium, and wide pH ranges are available in various strip lengths, and the strips are individually packaged and bar coded for convenient handling and sample tracking.
  • Improved Immobiline™ DryStrip gels provide increased stability, reproducibility, and protein spot definition
  • Better spot definition improves spot quantitation in data analysis
  • Immobiline™ DryStrip gels with matching IPG Buffer gives a high sample loading capacity and improves resolution
Immobiline™ DryStrip pH 3 - 11 NL covers most proteins found in prokaryotic and eukaryotic cells, while the narrow-range strips can be used when higher resolution is required. The pH gradients at the extreme ends of the pH scale are non linear (NL) to distribute the proteins evenly over the gel length and obtain maximal resolution.
Immobiline™ DryStrip gels are rehydrated with a matching IPG Buffer prior to use. Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of protein. Samples can also be loaded after rehydration.
Certifications: DIGE approved.
Ordering information: Instructions supplied with the gel. Suitable for industrial applications like analytical chemistry and research. For more detailed studies or when using greater protein loads, choose from four new overlapping strips covering the pH 3 to 11 range.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty