Would you like to log in again?
DeStreak Rehydration Reagent, Cytiva
Supplier: Cytiva
DeStreak
17-6003-18 17-6003-18 17-6003-19 17-6003-19
95056-444PK 236.35 USD InStock
CA95056-444L 95056-444 95056-442 CA95056-442L
DeStreak Rehydration Reagent, Cytiva
Electrophoresis Reagents Western Blotting Reagents
The appearance of streaks that distort 2-D electrophoresis maps is a common problem, occurring most frequently when running gels that contain regions greater than pH 7.0. Increased sample load, increased length of the IPG strip, or using a narrower pH gradient worsens the problem. Extra spots on 2-D gels, caused by non specific oxidation of proteins, is another difficulty encountered when running gels containing basic regions. Both streaking and non specific oxidation result in poorly resolved protein patterns and reduced reproducibility between electrophoresis runs.
  • Improves reproducibility and quality of 2-D gels by preventing streaking
  • Eliminates extra spots caused by non specific oxidation of proteins
  • Stabilises proteins throughout entire 2-D runs, ensuring simplified, well-resolved protein patterns
  • Highly suitable for both analytical and preparative gels
DeStreak™ Rehydration Solution effectively eliminates non specific oxidation by maintaining protein thiol groups in a single oxidation state, irrespective of sample load, pH range, or run length. Protein patterns are stabilised in IPG strips of any length and pH gradient ensuring the same stable and reproducible pattern in every analysis. The effectiveness of DeStreak™ Rehydration Solution is maintained at the high sample loads and long run times required for preparative gel electrophoresis.
DeStreak Rehydration Solution contains optimized concentrations of urea, thiourea, CHAPS, and DeStreak Reagent. The solution is ready for use after addition of the appropriate IPG buffer. DeStreak Reagent is also available separately in a 1-ml pack size.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty