Synonyms:
Thiocarbamide
, Sulfourea
, Sulphourea
RPN6301
RPN6301
95056-462EA
128.46
USD
InStock
CA95056-462L
95056-462
Thiourea
Thiourea
Thiourea is a chaotrope used together with urea in the IPG strip rehydration to solubilise and denature proteins, and improve the solubility of membrane proteins.