Synonyms:
2-Iodoacetamide
RPN6302
RPN6302
95056-464EA
196.25
USD
InStock
95056-464
CA95056-464L
Iodoacetamide, Proteomics Grade
Iodoacetamide
Used for the efficient alkylation of thiols while minimizing oxidation of competing thiol pairs in protein samples.
- Proteomic grade
- Easily soluble in cold and hot water
Ultrapure, proteomic-grade iodoacetamide is used for efficient alkylation of thiols while minimizing oxidation of the competing thiol pairs in protein samples.