Would you like to log in again?
Difco™ M17 Agar, BD Diagnostics
Supplier: BD Diagnostics
218571
95056-558EA 618.16 USD InStock
95056-558
Difco™ M17 Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Use for the cultivation and isolation of mesophilic lactic streptococci and for the selection of Streptococcus thermophilus from yogurt, cheese starters and other dairy products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty