Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ERP72 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPS-720-D SPS-720D ADI-SPS-720-F SPS-720F
95056-566EA 513 USD InStock
CA95056-564 95056-566 CA95056-566 95056-564
Anti-ERP72 Rabbit Polyclonal Antibody
Antibodies
Endoplasmic reticulum protein (ERp) 72, a 72 kDa protein localized to the endoplasmic reticulum, is a member of the Protein Disulfide Isomerase (PDI) family of proteins. Together with other ER resident proteins such as BiP, GRP94, and PDI, they serve as the molecular chaperones for proper folding of newly translocated and glycolsylated proteins such as Thyroglobulin (Tg) and Human chorionic gonadotropin (hCG)-beta.

  • For Immunofluorescence, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey and rabbit ERp72. Detects a band of ~72kDa by Western blot.

Type: Primary
Antigen: ERP72
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty