Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Coated VICRYL® (polyglactin 910) Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
VICRYL®
ETHJ340H ETHJ416H ETHJ869H ETHJ338H ETHJ546G ETHJ488G ETHJ495G ETHJ393H ETHJ310H ETHJ391H ETHJ214H ETHJ844G ETHJ315H ETHJ311H ETHJ339H ETHJ392H ETHJ218H ETHJ494G ETHJ337H ETHJ386H ETHJ303H ETHJ463G ETHJ451H ETHJ316H ETHJ409G ETHJ845G ETHJ317H ETHJ422H
95056-936BX 1059.9 USD InStock
95056-936 95056-938 95057-020 95056-878 95057-024 95057-022 95057-028 95057-126 95057-026 95057-018 95056-940 95056-942 95056-926 95056-928 95056-944 95057-030 95056-946 95057-134 95057-012 95057-132 95057-098 95057-010 95057-016 95057-136 95057-014 95057-008 95056-874 95056-876
Coated VICRYL® (polyglactin 910) Sutures, ETHICON, National Distribution and Contracting
Sutures
Absorbable sterile synthetic sutures feature exclusive coating technology for smooth passage through tissue, ease of knot tying, unsurpassed knot-sliding capabilities, and virtually no package memory for easier handling
Indicated for use in general soft tissue approximation and ligation, including use in ophthalmic and orthopedic procedures, but not for use in cardiovascular or neurological tissues. Not intended for use where extended approximation of tissue is required.
Ordering information: Each suture comes with one or two needles attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty