Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ETHILON® Nylon Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
ETHILON®
ETH1667H ETH699G ETH689G ETH662G ETH1669H ETH2815G ETH669H ETH662H ETH698H ETH7731G ETH1667G ETHG697G ETH697H ETH668G ETH661H
95057-082BX 263.07 USD InStock
95057-082 95057-080 95056-900 95057-074 95056-910 95056-965 95057-050 95057-072 95057-046 95057-078 95057-056 95057-032 95056-908 95056-880 95056-894
ETHILON® Nylon Sutures, ETHICON, National Distribution and Contracting
Sutures
Nonabsorbable, sterile, surgical, monofilament sutures composed of the long-chain, aliphatic polymers nylon 6 and nylon 6,6
Indicated for use in general soft tissue approximation and ligation, including use in cardiovascular, ophthalmic, and neurological procedures. Not intended for use where permanent retention of tensile strength is required.
Certifications: Sutures meet all requirements established by United States Pharmacopeia (USP) for nonabsorbable surgical sutures.
Ordering information: Each suture comes with one or two needles attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty