Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MONOCRYL™ (poliglecaprone 25) Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
MONOCRYL™
ETHY416H
95056-980BX 983.55 USD InStock
95056-980
MONOCRYL™ (poliglecaprone 25) Sutures, ETHICON, National Distribution and Contracting
Sutures
Absorbable sterile sutures are made from unique synthetic copolymers that provide unprecedented monofilament pliability and smooth passage through tissue
These sutures elicit lower tissue reaction than chromic gut suture. Ideal for subcuticular closure, ligation, skin repairs, bowel, peritoneum, uterus, and vaginal cuff. Indicated for use in general soft tissue approximation and ligation, but not for use in cardiovascular or neurological tissues, microsurgery, or ophthalmic surgery.
Ordering information: Each suture comes with a taper point needle attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty