Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ETHIBOND® EXCEL Polyester Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
ETHIBOND®
ETHX183H
95056-982BX 756.94 USD InStock
95056-982
ETHIBOND® EXCEL Polyester Sutures, ETHICON, National Distribution and Contracting
Sutures
Nonabsorbable, braided, sterile, surgical sutures are composed of polyethylene terephthalate
Prepared from fibers of high molecular weight, long-chain, linear polyesters having reccurent aromatic rings as an integral component. The highly adherent coating is a relatively nonreactive nonabsorbable compound which acts as a lubricant to mechanically improve the physical properties of the uncoated suture by improving handling qualities as contrasted to the braided, uncoated fiber. The suture is braided for optimal handling properties, and dyed green for enhanced visibility in the surgical field.
Ordering information: The sutures are SUTUPAK® pre-cut sutures and do not come with needles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty