Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Chromic Gut Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
ETHU203H ETHS111H
95056-962BX 1321.64 USD InStock
95056-962 95056-984
Chromic Gut Sutures, ETHICON, National Distribution and Contracting
Sutures
Absorbable, sterile surgical sutures are composed of purified connective tissue (mostly collagen) derived from either the serosal layer of bovine intestines or the submucosal fibrous layer of ovine intestines
Chromic gut is processed to provide greater resistance to absorption. Surgical gut is packaged in tubing fluid. Indicated for use in general soft tissue approximation and ligation, including use in ophthalmic procedures, but not for use in cardiovascular and neurological tissues. Not for use where extended approximation of tissue is required. The use of this suture is contraindicated in patients with known sensitivities or allergies to collagen or chromium, as gut is a collagen based material, and chromic gut is treated with chromic salt solutions.
Certifications: Surgical gut suture meets all requirements established by the US Pharmacopeia (USP) for absorbable surgical sutures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty