Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PERMA-HAND™ Silk Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
PERMA-HAND™
ETHK890H ETH737G ETHK831H ETHA185H ETH640G ETHLA53G ETH443H ETH677G ETHK891H ETH734G ETHK571H ETHK873H ETH683G ETHM104T ETHN266H ETHK809H ETHA182H ETH1679H ETH768G ETH706G ETH682G ETHK880H ETH736G ETH639G ETHN267H ETH678G ETHK833H ETH632G ETHK889H ETHK870H ETHLA54G ETH675H ETHSA64H ETHK802H ETH7734G ETHK834H ETH624H ETHK882H ETH707G ETHK872H ETH685G ETHK952H ETHLA55G
95057-060BX 232.8 USD InStock
95057-060 95056-914 95056-916 95057-064 95057-086 95056-954 95056-998 95057-062 95056-912 95056-956 95057-002 95057-068 95057-088 95057-000 95057-006 95057-004 95057-048 95056-960 95056-948 95057-092 95057-070 95057-090 95056-988 95056-922 95056-966 95057-052 95057-096 95056-968 95057-094 95057-034 95057-054 95057-058 95056-890 95056-896 95056-950 95056-972 95056-994 95056-996 95056-952 95056-870 95056-990 95056-872 95056-992
PERMA-HAND™ Silk Sutures, ETHICON, National Distribution and Contracting
Sutures
Nonabsorbable, sterile, surgical sutures are composed of an organic protein called fibroin, which is derived from the domesticated species Bombyx mori (B
mori) of the family Bombycidae. Indicated for use in general soft tissue approximation and ligation, including use in cardiovascular, ophthalmic and neurological procedures. Contraindicated in patients with known sensitivities or allergies to silk. Not recommended where permanent retention of tensile strength is required. Sutures are processed to remove the natural waxes and gums, dyed black, and coated with a special wax mixture.
Certifications: Sutures meet requirements established by the United States Pharmacopeia (USP) for nonabsorbable surgical sutures.
Ordering information: Each suture comes with one or two needles attached, except for LIGAPAK® dispensing reels and SUTUPAK® pre-cut sutures, which do not come with needles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty