Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PROLENE™ Polypropylene Sutures, ETHICON, National Distribution and Contracting
Supplier: National Distribution and Contracting
PROLENE™
ETH8683G ETH8889H ETH8831H ETH8711H ETH8557H ETH8862H ETH8661G ETH8834H ETHM8718 ETH8807H ETH8833H ETH1713G ETH8714H ETH8699G ETH8718H ETH8881H
95057-130BX 6234.08 USD InStock
95057-130 95056-888 95057-040 95057-044 95057-120 95057-076 95057-116 95056-918 95057-038 95057-036 95057-124 95057-118 95056-884 95056-920 95056-986 95056-886
PROLENE™ Polypropylene Sutures, ETHICON, National Distribution and Contracting
Sutures
Nonabsorbable, sterile surgical sutures composed of an isotactic crystalline stereoisomer of polypropylene, a synthetic linear polyolefin
Indicated for use in general soft tissue approximation and ligation, including use in cardiovascular, ophthalmic, and neurological procedures.
Certifications: Sutures meet all requirements established by United States Pharmacopeia (USP) for nonabsorbable surgical sutures.
Ordering information: Each suture comes with one or two needles attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty