Would you like to log in again?
VACUETTE® Glycolytic Inhibitor Tubes, Greiner Bio-One
Supplier: Greiner bio-one
Vacuette®
454238 456020 454297
95057-185CS 298.1 USD InStock
95057-185 95057-189 95057-187
VACUETTE® Glycolytic Inhibitor Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Ideal for use in glucose and lactate determinations, tubes contain an anticoagulant (potassium oxalate) and a stabilizer (sodium fluoride).
  • PET plastic construction is virtually unbreakable
  • Color-coded rings in caps
  • Tubes contain anticoagulant and stabilizer
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with grey non-ridged pull caps that feature an exterior color-coded ring to indicate the draw volume. A black ring indicates standard tubes and a white ring indicates a tube designed to accommodate pediatric draw.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty