Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VACUETTE® Blood Collection Needles, Greiner Bio-One
Supplier: Greiner bio-one
Vacuette®
450075 451048 450071 450072 450061 450070 450077 451069 450073 451047 450062 450076
76302-842CS 163.71 USD InStock
76302-842 95057-205 95057-195 76359-810 95057-353 95057-199 76302-848 95057-197 95057-203 76302-844 76359-812 95057-201
VACUETTE® Blood Collection Needles, Greiner Bio-One
Syringe Needles
These needles are ideal for use in routine blood collection procedures.
  • Color-coded for simple identification of needle gauge
  • Designed for single use
  • Single-sample needles for veterinary use only
They are designed exclusively for single use, and feature color-coded protective needle caps to simplify identification of the needle gauge. Pink caps correspond to 18G needles, yellow caps to 20G, green caps to 21G, and black caps to 22G. Available in two lengths, 25.4 or 38.1 mm (1 or 11/2"). Single-sample needles are designed for veterinary use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty