Would you like to log in again?
MiniCollect® Serum and Plasma Tubes, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MiniCollect®
450472 450477 450470 450475 450479 450480
95057-291CS 1364.94 USD InStock
95057-291 95057-295 95057-299 95057-286 95057-297 95057-285
MiniCollect® Serum and Plasma Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Tubes are suitable for collection, transport, and processing of capillary blood samples from infants, toddlers, children, geriatric patients, and patients in intensive care.
  • May be used to test clinical chemistry parameters
  • Stable gel barrier for transport
  • Available in a variety of formats for specific uses
Serum and plasma tubes may be used for testing parameters in clinical chemistry
The EDTA tubes may be used for testing parameters in hematology. Carrier tubes are designed to ensure compatibility with most common centrifuges and laboratory instruments. The blood collection capillary and funnel are used to simplify blood transfer.
Tubes with a gel separator contain an inert, acrylic barrier in the base of the tube with a specific gravity that lies between the clot/cells and serum/plasma. During centrifugation, the gel will shift to form a stable barrier between the clot/cells and the serum/plasma. The barrier remains stable for transportation. These application-specific tubes are available in a variety of fill volumes.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty