Would you like to log in again?
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vacuette®
456292P 454228P 455092 454236 454204 454243P 456089 456073P 455071P 454067P 455099P 456018P
95057-383CS 403.62 USD InStock
95057-383 95057-371 95057-393 95057-381 95057-391 95057-387 95057-365 95057-397 95057-385 95057-369 95057-389 95057-377
VACUETTE® Serum Clot Activator Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Tubes are designed for routine use in immunohematology testing and viral marker testing in screening and clinical laboratories.
  • Gel forms stable barrier between serum and blood cells
  • Tubes are effective in therapeutic drug monitoring testing
The inner wall of each serum tube is coated with microscopic silica particles which activate the coagulation process when tubes are inverted
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow/gold ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.
Tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells, separating the serum from fibrin and cells. These tubes are particularly effective in therapeutic drug monitoring testing. Standard tubes are also available with no additive.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty