Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Giemsa's stain solution for hematology, used in parasitology

Supplier: AZER SCIENTIFIC INC
Synonyms: Giemsa's azur-eosin-methylene blueAzure mixture sicc. Giemsa stainAzur-Eosin-Methylene blue acc. Giemsa

ES915 ES915 ES915-16 ES915-16 ES917
95057-502EA 111.45 USD InStock
CA95057-502 95057-502 CA89130-204 95057-506 89130-204
Giemsa's stain solution for hematology, used in parasitology
Giemsa's stain
Giemsa stain is a classical blood film stain for peripheral blood smear and bone marrow specimens.

  • For the visualization of blood parasites

Erythrocytes stain pink, platelets show a light pale pink, lymphocyte and monocyte cytoplasm stain pale blue and leukocyte nuclear chromatin stains magenta.

Store at controlled room temperature (15-30°C). Keep bottle tightly closed when not in use. Do not use after expiration date specified on bottle label. Excessive precipitation in reagent is an indication of degradation


Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty