Would you like to log in again?
Wright-Giemsa stain solution
Supplier: AZER SCIENTIFIC INC
Synonyms: Wright-Giemsa Solution
ES923 ES921
95057-514EA 65.46 USD InStock
95057-514 95057-518
Wright-Giemsa stain solution
Wright-Giemsa stain (solutions & kits)
Wright-Giemsa stain is a modified Romanowsky stain used for staining the cellular elements in blood and bone marrow
  • Provides excellent differential staining results
  • Stains RBC inclusion bodies, parasites and other organisms in blood smears
  • Sutiable for automated or manual staining methods
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty