Would you like to log in again?
Phosphate Buffer Solutions for Hematology, Azer Scientific
Supplier: Azer Scientific
ES927 ES928 ES9275
89129-900CS 255.99 USD InStock
89129-900 95057-522 95057-520
Phosphate Buffer Solutions for Hematology, Azer Scientific
Buffers Biological Buffers
Low molarity phosphate buffers optimized for use with Azer Scientific Wright and Wright-Giemsa stain solutions.
  • Suitable for use with any Wright, Wright-Giemsa, or Giemsa stain solutions
  • Low molar strength formula extends the life of stain dilutions
  • Available in pH range from 6.6-7.0 to achieve desired stain intensity and colors
Ready to use phosphate buffer optimized for use in manual and automated hematology stain protocols.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty