Would you like to log in again?
Proteinase K
Supplier: G-Biosciences
786-044 786-043 786-043 786-044
95057-534EA 879.39 USD InStock
CA95057-534 CA95057-532 95057-534 95057-532
Proteinase K
Enzymes
Proteinase K (also protease K, endopeptidase K, peptidase K or Tritirachium alkaline phosphatase) (EC 3.4.21.64) is a non-specifc, broad spectrum serine protease that is isolated from the saprophytic fungus Tritirachium album.
Proteinase K is routinely used for the purification of target material from contaminating proteins, for the isolation of RNA and DNA and to inactivate other protein enzymatic activities. Proteinase K exhibits high activity in the presence of SDS, urea and EDTA. This allows the use of EDTA in the purification of DNA or RNA to inhibit calcium dependent nucleases.
The dry, lyophilized powder is highly soluble (>50mg/mL), however stock solutions are normally 20mg/mL. The proteinase K is completely RNase and DNase free and has a specific activity of >30 units/mg at 37°C. The recommended storage buffer is 20mM Tris-HCl (pH7.4), 1mM CaCl2, 50% glycerol. Proteinase K is stable for 1 year in this storage buffer, recommended storage is -20°C.
Ordering information: Accessories not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty