Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, 2D-Detergent™, purified

Supplier: G-Biosciences
Synonyms: (p-tert-Octylphenoxy)polyethoxyethanolO-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanolOctoxynolPolyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether

DG907 DG908 DG908 DG907
95057-624EA 441.42 USD InStock
CA95057-624 95057-624 CA95057-622 95057-622
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, 2D-Detergent™, purified
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds

2D-Detergent™ solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 15µS conductivity. These detergents are offered as 10% aqueous solutions, sealed under inert gas and are suitable for all protein applications, including 2D-electrophoresis. These non-ionic detergents are suitable for isolating membrane-protein complexes. The aldehyde and peroxide levels are <50µM and <10µM respectively with a conductivity of <15µS.

The proteins that are isolated with these detergents are highly susceptible to contaminating peroxides and carbonyls. The peroxides will oxidize proteins and the carbonyl groups will form Schiff's bases with the proteins that interfere with a protein's structure.

Ordering information: Accessories not included.


MDL Number: MFCD00132505
CAS Number: 9002-93-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty