Would you like to log in again?
Adenine Deaminase
Supplier: G-Biosciences
786-428 786-428
95057-674EA 578.6 USD InStock
CA95057-674 95057-674
Adenine Deaminase
Enzymes
Adenine Deaminase converts Adenine to Hypoxanthine.
Adenine Deaminase is a highly purified recombinant enzyme protein supplied at a 2 µM concentration.
Ordering information: Accessories available.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty