Would you like to log in again?
Deoxynucleotides (dNTPs), G-Biosciences
Supplier: G-Biosciences
786-468 786-465 786-467 786-468 786-467 786-443 786-460 786-460 786-466 786-443 786-466 786-465 786-442 786-442
95057-688EA 226.33 USD InStock
CA71003-178 95057-688 95057-676 71003-180 CA95057-678 71003-182 CA95057-688 CA71003-180 95057-678 CA95057-676 71003-178 71003-184 CA71003-182 CA71003-184
Deoxynucleotides (dNTPs), G-Biosciences
Nucleic Acid Reagents Nucleotides
G-Biosciences' Deoxynucleotides (dNTPs) consist of aqueous solutions at pH7.0 of dATP, dCTP, dGTP or dTTP in 100 mM concentration, each supplied in individual vials
In addition, G-Biosciences offers a dNTP Mix is a ready-to-use, aqueous solution at pH7.0, that contains dATP, dCTP, dGTP and dTTP, each at a final concentration of 10 mM. The dNTP Mix has been functionally tested in PCR with Taq and Pfu DNA polymerases. The mix is endo/exoribonuclease-, ribonuclease- and phosphatase-free. The mix reduces the number of pipeting steps and the risk of errors.
G-Biosciences' deoxynucleotides are for research use only (not for clinical use) in PCR, long PCR, RT-PCR, cDNA synthesis, primer extension, DNA sequencing and DNA labeling.
Although the nucleotides are provided separately, G-Biosciences also offers a dNTP Set, for maximum flexibility in the preparation of reaction mixes for different applications.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty