Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAMK2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-CA002D KAP-CA002F KAP-CA002-F KAP-CA002-D
95057-952EA 593 USD InStock
CA95057-952 CA95057-950 95057-952 95057-950
Anti-CAMK2 Rabbit Polyclonal Antibody
Antibodies
CaMKII (calmodulin-dependent kinase II) is an enzyme which is activated by increases in intracellular Ca2+ ion concentration and it has been proposed to be pivotal in regulating synaptic strength and maturation of synapses during development.  This process is thought to be critical in memory and learning and in establishing the specificity of synaptic connections. There are over two dozen alternative splice variants of CaMKII which are encoded by four genes, a, b, g, and d with apparent molecular masses of 50-60 kDa.  CaMKII is widely distributed in many tissues, but is highly expressed in brain. Several studies demonstrate that CaMKII is a multifunctional enzyme which modulates the synaptic strength by binding to a subunit of NMDA receptors and promoting the phosphorylation of this NMDA receptor subunit.  CaMKII also regulates DLG localization at synapses by co-localization with DLG in the same protein complex.  Experimental data suggest that CaMKII is critically involved in the development of morphine tolerance as well as dependence and inhibition of this enzyme may have some therapeutic benefit in the treatment of opiate tolerance and dependence.  It also has been demonstrated that d CaMKII isozyme is down-regulated in human tumor cells indicating a role for d CaMKII isozymes in cellular differentiation.  Changes in d CaMKII isozyme expression pattern in human hearts during heart failure suggest that CaMKII is important for regulation of heart function.  This immunoaffinity purified antibody detects proteins of 50-60 kDa, corresponding to apparent molecular mass of CaMKII isoforms on SDS-PAGE immunoblots, in samples from human, mouse, rat, bovine, hamster, guinea pig, chicken and rabbit origins. Recombinant rat calmodulin-dependent protein kinase II a subunits expressed in sf9 insect cells are also detected. As the sequences of the rat a, d, and g isoforms are conserved over this amino terminus region, this antibody is expected to recognize the d, g and a isoforms. This antibody is not expected to cross-react with the b isoform. Proteins of unknown identity, ~40, 45 and 90 kDa, may be detected on immunoblot analysis with some lysates.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, hamster, guinea pig, chicken and rabbit CaMKII. Detects a band of ~50-60kDa by Western blot.

Type: Primary
Antigen: CAMK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty