Would you like to log in again?
Wash Solution Concentrate, 20X, SeraCare
Supplier: SeraCare
51-50-0011 5150-0008
95059-134EA 200.02 USD InStock
95059-134 95059-132
Wash Solution Concentrate, 20X, SeraCare
Buffers Electrophoresis Buffers Washing Buffers
Convenient 20X concentrate containing imidazole-buffered saline and Tween® 20
  • Suitable for ELISA, blotting, and immunohistochemistry applications
This is used a general immunoassay wash buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty