Would you like to log in again?
Phosphatase Wash Solution 5X, SeraCare
Supplier: SeraCare
5960-0018
95059-140EA 236.17 USD InStock
95059-140
Phosphatase Wash Solution 5X, SeraCare
Buffers Electrophoresis Buffers Washing Buffers
Wash solution for use with alkaline phosphatase based detection systems on membranes.
Phosphatase wash solution (5X) is formulated to effectively remove unbound phosphatase conjugate and improve assay performance. It is intended for use with phosphatase assay buffer and CDP-star chemiluminescent substrate in procedures such as western, southern, northern, and dot blotting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty