Would you like to log in again?
Phosphate Buffered Saline (PBS), 10X, SeraCare
Supplier: SeraCare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5460-0022 5460-0023
95059-254EA 299.56 USD InStock
95059-254 95059-252
Phosphate Buffered Saline (PBS), 10X, SeraCare
Buffers Assay Buffers
10X Phosphate-buffered Saline (PBS) is ideal for use in sample preparation and as a wash buffer in general immunoassay applications.
  • Conductivity controlled
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty