Would you like to log in again?
10X TBS Wash Buffer, with Tween 20, Sera Care
Supplier: SeraCare
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
5460-0044 5460-0040 5460-0041 5460-0043
95059-278EA 293.91 USD InStock
95059-278 95059-276 95059-274 95059-272
10X TBS Wash Buffer, with Tween 20, Sera Care
Buffers Assay Buffers
10X Tris-buffered Saline (TBS) with Tween 20 is used as a common western blot wash solution and general wash buffer in immunoassay applications
Formulation of TBST with higher concentration of Tween 20 provides a more stringent wash than standard TBST formulations.
Tween 20, a non-ionic detergent, reduces non-specific binding and protein-protein interaction during the wash step in immunoassay procedures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty