Would you like to log in again?
Protein Detector LumiGLO Western Blot Kit, KPL
Supplier: KPL
5410-0009
95059-302EA 884.19 USD InStock
95059-302
Protein Detector LumiGLO Western Blot Kit, KPL
Stains Electrophoresis Stains
The Protein Detector LumiGLO Western Blot Kit is designed for low picogram detection of proteins captured on immunoblots
This kit is a fully optimized system that delivers high signal with minimal background. The ready-to-use reagents enable convenient assay preparation.
The combination of a highly specific, stable liquid conjugate and a sensitive, chemiluminescent substrate allows rapid and accurate identification of samples. All solutions required for blocking and washing the membrane and for diluting antibodies are provided. For added flexibility, affinity purified anti-mouse and anti-rabbit secondary antibodies conjugated with horseradish peroxidase are provided in stabilized, liquid form for quick dilution. LumiGLO Chemiluminescent Substrate is provided as a stable two component solution, which emits light when reacted with the HRP-labeled antibody.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty