Would you like to log in again?
LumiGLO Peroxidase Chemiluminescent Substrate
Supplier: KPL
5430-0041 5430-0042 5430-0040
95059-320EA 152.86 USD InStock
95059-320 95059-318 95059-316
LumiGLO Peroxidase Chemiluminescent Substrate
Electrophoresis Reagents Western Blotting Reagents
LumiGLO Peroxidase Chemiluminescent substrate is an all-purpose, luminol-based substrate designed for the rapid and sensitive detection of peroxidase labeled-conjugates in Western blotting and ELISA
It enables low-picogram detection and increased sensitivity over colorimetric substrates in both applications. In blotting applications, permanent results are recorded on X-ray film. Long signal duration allows for multiple stripping and reprobing of blots. In ELISA, positive reactions are read and rapidly detected using a luminometer.
It is supplied as a 2-component, mix and use solution.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty