Would you like to log in again?
Protein Detector BCIP/NBT Western Blotting Kit, KPL
Supplier: KPL
5410-0013
95059-338EA 815.02 USD InStock
95059-338
Protein Detector BCIP/NBT Western Blotting Kit, KPL
Stains Electrophoresis Stains
The Protein Detector BCIP/NBT Western Blot Kit is designed for the colorimetric detection of proteins immobilized on membranes through either electrophoresis or dot blotting
The Protein Detector BCIP/NBT Western Blotting Kit is ideal for routine colorimetric detection of overexpressed protein. It provides sensitivity in the 500 picogram range, and contains ready-to-use reagents for quick and convenient assay preparation. BCIP/NBT Substrate is stable for one year. Both anti-rabbit and anti-mouse antibodies are supplied for versatility.
The combination of highly specific alkaline phosphatase-labeled mouse and rabbit secondary antibodies with a sensitive colorimetric substrate allows rapid and accurate identification of samples. All solutions required for blocking and washing the membrane and for diluting antibodies are provided. For added flexibility, affinity purified anti-mouse and anti-rabbit secondary antibodies conjugated with alkaline phosphatase are provided in stabilized, liquid form for quick dilution. The BCIP/NBT substrate is provided as a convenient, ready-to-use solution, which forms an insoluble purple precipitate when reacted with the AP-labeled antibody.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty