Would you like to log in again?
Protein Detector™ Phosphatase ELISA Kits, Lgc Clinical Diagnostics, Inc
Supplier: LGC Clinical Diagnostics
Protein Detector™
5110-0012
95059-364EA 545.85 USD InStock
95059-364
Protein Detector™ Phosphatase ELISA Kits, Lgc Clinical Diagnostics, Inc
Assays ELISAs
AP ELISA Kits offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection
Kit contains Coating Solution Concentrate; BSA Diluent/Blocking Solution Concentrate; Wash Solution Concentrate; 50% Glycerol Solution; AP labeled Goat Anti-Human IgG (H+L); AP labeled Goat Anti-Rabbit IgG (H+L); BluePhos Phosphatase Substrate System; and APstop Solution Concentrate.
Highly sensitive reagents are optimized for antigen detection in picogram quantities. The wash and block solutions ensure low background. This kit can be used for direct and indirect detection.
These kits provide pretested components and suggested protocols to facilitate the development of ELISA procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty