Would you like to log in again?
Mag-Bind SeqDTR, Omega Bio-Tek
Supplier: Omega Bio-Tek
Mag-Bind®
M1300-08 M1300-05 M1300-05 M1300-50 M1300-50 M1300-08
95059-686EA 1575.57 USD InStock
CA95059-684 95059-686 95059-684 CA95059-688 CA95059-686 95059-688
Mag-Bind SeqDTR, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Omega Bio-teks Mag-Bind SeqDTR is designed to effective and reliable removal of unincorporated terminators from sequencing reaction. The system combines Omega Bio-teks proprietary chemistry with the reversible nucleic acid-binding properties of paramagnetic beads to eliminate excess nucleotides, primers, salts, and unicorporated dye terminators.
  • Long Phred 20 read lengths averaging over 800 bps
  • Works with automated liquid handlers
  • Can be used in 96 or 384 format
This kit is designed for both manual and fully automated purification of sequencing products, The chemistry has been successfully automated on liquid handlers such has Beckman Coulter’s Biomek FX/NX, and Hamilton STAR/Starlet. Mag-Bind SEQ DTR high recovery rate allows for decreased usage of ABI Big Dye chemistry, normally 1/16 or 1/32 reactions can be performed without a decrease in signal strength when compared to sephadex based clean-up
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty