Would you like to log in again?
Protocol™ Parasitology Concentrate Systems, Thermo Scientific
Supplier: Thermo Scientific
Protocol™
23-005-51 23-005-50
95059-792PK 661.29 USD InStock
95059-792 95059-770
Protocol™ Parasitology Concentrate Systems, Thermo Scientific
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection
These complete systems are for concentrating and recovering helminth eggs, larvae, and protozoan cysts from stool specimens
Proper use optimizes isolation and identification while offering an easy-to-use processing system that minimizes specimen handling. Specimens collected and transported in 10% formalin, SAF, or empty vials may be used with these systems
Ordering information: The 50mL concentration system contains 120 filtering devices, 120 conical centrifuge tubes and screw caps, and 30mL surfactant. The 15mL concentrate system contains 50 filtering devices, 50 disposable 15mL centrifuge tubes and caps, Reagent A (surfactant), and Reagent B (ethyl acetate).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty