Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Supplier: Corning
Corning®
354784 356784 354784 356784
95059-902CS 78.81 USD InStock
CA11006-216 95059-902 95059-922 CA11006-196
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Culture Dishes Cell Culture Dishes
PS. Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A proprietary thin-film coating technology produces a uniform, functionalised surface that provides a highly controlled environment for cell culture applications.
  • Available as amine (positively charged) or carboxyl (negatively charged) surface chemistries
  • Optimised for culture in low serum or no serum conditions
  • Enhanced freeze-thaw recovery of cryopreserved cells
  • Improved differentiation of cells
  • Culture of many cell lines and primary cells supported, including primary neuronal cells and stem cells
A proprietary thin-film coating technology produces a uniform, functionalized surface that provides a highly controlled environment for cell culture applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty