Would you like to log in again?
Temperature Controller, Stoelting
Supplier: Stoelting
50322 50321 50320
95060-048EA 1899.52 USD InStock
95060-048 95060-050 95060-052
Temperature Controller, Stoelting
Controllers Temperature Controllers
These general purpose temperature controller (TC) provides accurate temperature control in a variety of laboratory applications. Combined with a heat lamp or heating pad, the TC Unit may be used to maintain the body temperature of anesthetized animals during surgery or stereotaxic procedures. Small animal cages are easily warmed with a heat lamp.
  • Durable, baked enamel painted aluminum housing
  • Adjustable front feet allow the user to raise the front of controller to provide a more convenient viewing angle
TCAC: This version controls only powered heating or cooling devices such as a heat lamp
TCDC: This model provides a low voltage DC output for electrically sensitive applications where€ frequencies might interfere with other monitoring devices. It comes standard with a built in DC power supply designed to operate with our Heat Plate for small rodents.
TCACDC: This is a combination version which includes both an outlet for controlling primarily resistance loads such as heat lamps and a low-voltage, low noise DC output for heat plate. can control a variety of different loads.
The TC is simple to set-up; connect the feedback sensor Type T thermocouple probe to the animal and to the back of the controller, connect the heating plate or lamp to the appropriate outlet on the rear of the controller. Adjust the set temperature as required (lower green display) according to the instruction label on top of the instrument. The set temperature is stored in memory after the unit is switched off.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty